Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bspry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138653.1 | NP_619594.1 | 3 - 4 | 701 | 71 | |
GenBank mRNA | AJ276690.1 | - | 3 - 4 | 701 | 71 | |
AK035588.1 | - | 3 - 4 | 707 | 71 | ||
AK160930.1 | - | 3 - 4 | 707 | 71 | ||
BC003992.1 | - | 1 - 2 | 244 | 71 | ||
BC139079.1 | - | 3 - 4 | 717 | 8862101 | 71 | |
BC139080.1 | - | 3 - 4 | 717 | 8862102 | 71 | |
BC145535.1 | - | 3 - 4 | 717 | 9054098 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |