Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139146.2 | NP_631885.1 | 8 - 9 | 1674 | 96 | |
GenBank mRNA | AF319623.1 | - | 8 - 9 | 1674 | 96 | |
AJ606045.1 | - | 7 - 8 | 1392 | 96 | ||
AK129270.1 | - | 7 - 8 | 2665 | 96 | ||
BC138625.1 | - | 7 - 8 | 1425 | 8861645 | 96 | |
BC138626.1 | - | 7 - 8 | 1425 | 8861646 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |