Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144522.5 | NP_653105.3 | 4 - 5 | 1334 | 72 | |
GenBank mRNA | AF285112.1 | - | 4 - 5 | 692 | 72 | |
AK047320.1 | - | 4 - 5 | 1335 | 72 | ||
AK047327.1 | - | 4 - 5 | 1335 | 72 | ||
BC072576.1 | - | 4 - 5 | 877 | 6401332 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |