Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199186.1 | NP_001186115.1 | 2 - 3 | 179 | 65 | |
NM_152808.3 | NP_690021.1 | 2 - 3 | 181 | 65 | ||
GenBank mRNA | AK041533.1 | - | 2 - 3 | 138 | 65 | |
AK043924.1 | - | 2 - 3 | 151 | 65 | ||
AK048648.1 | - | 2 - 3 | 166 | 65 | ||
AK088553.1 | - | 2 - 3 | 128 | 65 | ||
AK153992.1 | - | 2 - 3 | 160 | 65 | ||
AK166695.1 | - | 2 - 3 | 182 | 65 | ||
BC023763.1 | - | 2 - 3 | 164 | 65 | ||
BC031535.1 | - | 2 - 3 | 157 | 3991609 | 65 | |
BY339157.1 | - | 2 - 3 | 181 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |