Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291903.1 | NP_001278832.1 | 2 - 3 | 2295 | 61 | |
NM_009623.2 | NP_033753.2 | 2 - 3 | 2295 | 61 | ||
GenBank mRNA | AK138244.1 | - | 2 - 3 | 2296 | 61 | |
AK163443.1 | - | 2 - 3 | 2271 | 61 | ||
BC059010.1 | - | 2 - 3 | 1393 | 61 | ||
BC094231.1 | - | 2 - 3 | 1733 | 61 | ||
U85021.1 | - | 2 - 3 | 2155 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |