Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009710.4 | NP_033840.2 | 3 - 4 | 926 | 109 | |
GenBank mRNA | AK003107.1 | - | 3 - 4 | 927 | 109 | |
AK009284.1 | - | 3 - 4 | 255 | 109 | ||
AK009902.1 | - | 927 | 109 | |||
BC063760.1 | - | 3 - 4 | 907 | 30137479 | 109 | |
BC132492.1 | - | 3 - 4 | 914 | 40130769 | 109 | |
BC132496.1 | - | 3 - 4 | 914 | 40130773 | 109 | |
X95842.1 | - | 2 - 3 | 888 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |