Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009902.1 | - | 1477 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |