Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164099.2 | NP_001157571.1 | 4 - 5 | 781 | 102 | |
NM_001164100.2 | NP_001157572.1 | 4 - 5 | 843 | 102 | ||
NM_001164101.2 | NP_001157573.1 | 4 - 5 | 781 | 102 | ||
NM_001277100.1 | NP_001264029.1 | 4 - 5 | 843 | 102 | ||
NM_013758.4 | NP_038786.2 | 5 - 6 | 593 | 102 | ||
GenBank mRNA | AF100424.1 | - | 3 - 4 | 512 | 102 | |
AF100425.1 | - | 3 - 4 | 512 | 102 | ||
AK019491.1 | - | 4 - 5 | 722 | 102 | ||
AK028481.1 | - | 4 - 5 | 686 | 102 | ||
AK035151.1 | - | 4 - 5 | 610 | 102 | ||
AK083939.1 | - | 4 - 5 | 684 | 102 | ||
AK144174.1 | - | 4 - 5 | 787 | 102 | ||
BC029196.1 | - | 4 - 5 | 646 | 102 | ||
BC037116.1 | - | 5 - 6 | 604 | 4506748 | 102 | |
BC060097.1 | - | 4 - 5 | 625 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |