Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164099.2 | NP_001157571.1 | 9 - 10 | 1437 | 69 | |
NM_001164100.2 | NP_001157572.1 | 9 - 10 | 1499 | 69 | ||
NM_001164101.2 | NP_001157573.1 | 9 - 10 | 1437 | 69 | ||
NM_001277100.1 | NP_001264029.1 | 9 - 10 | 1499 | 69 | ||
NM_013758.4 | NP_038786.2 | 10 - 11 | 1249 | 69 | ||
GenBank mRNA | AF100424.1 | - | 8 - 9 | 1168 | 69 | |
AF100425.1 | - | 8 - 9 | 1168 | 69 | ||
AK019491.1 | - | 9 - 10 | 1378 | 69 | ||
AK028481.1 | - | 9 - 10 | 1342 | 69 | ||
AK035151.1 | - | 9 - 10 | 1266 | 69 | ||
AK083939.1 | - | 9 - 10 | 1340 | 69 | ||
AK144174.1 | - | 9 - 10 | 1443 | 69 | ||
BC004032.1 | - | 9 - 10 | 1216 | 69 | ||
BC004746.1 | - | 2 - 3 | 247 | 69 | ||
BC029196.1 | - | 9 - 10 | 1302 | 69 | ||
BC037116.1 | - | 10 - 11 | 1260 | 4506748 | 69 | |
BC060097.1 | - | 9 - 10 | 1281 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |