Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020586.2 | NP_065611.1 | 2 - 3 | 1035 | 75 | |
GenBank mRNA | AB041580.1 | - | 2 - 3 | 830 | 75 | |
AK019862.1 | - | 2 - 3 | 980 | 75 | ||
AK135935.1 | - | 2 - 3 | 1036 | 75 | ||
BC029691.1 | - | 2 - 3 | 253 | 5375603 | 75 | |
BC043693.1 | - | 2 - 3 | 1074 | 5150485 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |