Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020586.2 | NP_065611.1 | 6 - 7 | 1758 | 112 | |
GenBank mRNA | AB041580.1 | - | 6 - 7 | 1553 | 112 | |
AK019862.1 | - | 6 - 7 | 1703 | 112 | ||
AK135935.1 | - | 6 - 7 | 1759 | 112 | ||
BC029691.1 | - | 6 - 7 | 976 | 5375603 | 112 | |
BC043693.1 | - | 6 - 7 | 1797 | 5150485 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |