Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020622.2 | NP_065647.1 | 4 - 5 | 438 | 101 | |
GenBank mRNA | AF360358.1 | - | 4 - 5 | 439 | 101 | |
AF494380.1 | - | 4 - 5 | 343 | 101 | ||
AK018571.1 | - | 4 - 5 | 408 | 101 | ||
BC048949.1 | - | 4 - 5 | 385 | 6474753 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |