Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042592.2 | NP_001036057.1 | 1 - 2 | 475 | 110 | |
NM_025549.3 | NP_079825.2 | 1 - 2 | 475 | 110 | ||
GenBank mRNA | AK010022.1 | - | 1 - 2 | 441 | 110 | |
AK037198.1 | - | 1 - 2 | 442 | 110 | ||
BC017528.1 | - | 1 - 2 | 425 | 4922348 | 110 | |
BY738896.1 | - | 1 - 2 | 476 | 110 | ||
DQ861996.1 | - | 1 - 2 | 279 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |