Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025692.3 | NP_079968.2 | 2 - 3 | 375 | 77 | |
GenBank mRNA | AK017787.1 | - | 2 - 3 | 375 | 77 | |
AK077825.1 | - | 2 - 3 | 357 | 77 | ||
AK151823.1 | - | 2 - 3 | 332 | 77 | ||
AK152029.1 | - | 2 - 3 | 370 | 77 | ||
AK168907.1 | - | 2 - 3 | 369 | 77 | ||
BC019764.1 | - | 2 - 3 | 252 | 3989119 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |