Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025693.4 | NP_079969.1 | 1 - 2 | 189 | 119 | |
GenBank mRNA | AK007685.1 | - | 1 - 2 | 184 | 119 | |
AK160292.1 | - | 1 - 2 | 109 | 119 | ||
BB853849.1 | - | 1 - 2 | 189 | 119 | ||
BC019770.1 | - | 1 - 2 | 144 | 5124769 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |