Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025819.4 | NP_080095.3 | 11 - 12 | 1732 | 78 | |
GenBank mRNA | AK004803.2 | - | 11 - 12 | 1679 | 78 | |
BC055064.1 | - | 11 - 12 | 1733 | 6528686 | 78 | |
BC060204.1 | - | 11 - 12 | 1672 | 6855442 | 78 | |
CV563132.1 | - | 1 - 2 | 76 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |