Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025914.2 | NP_080190.1 | 7 - 8 | 719 | 150 | |
GenBank mRNA | AK008409.1 | - | 7 - 8 | 720 | 150 | |
AK088571.1 | - | 7 - 8 | 705 | 150 | ||
BC062137.1 | - | 7 - 8 | 696 | 6509757 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |