Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988730 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988729 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988731 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988732 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
475610 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897739 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948874 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
361115 |
AK132558.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
361115 |
AK162468.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
361115 |
NM_001033416.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
444732 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
957725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
337591 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9679 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496054 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
89657 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485178 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760865 |
AK004847.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760865 |
AK146266.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760865 |
AK167031.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760865 |
NM_023270.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107266 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P122749 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P117883 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988760 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988714 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988712 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988747 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988719 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988731 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988753 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988734 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988741 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988750 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988743 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988749 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988751 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988717 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988720 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28988735 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P122749 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P117883 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07854308 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07854288 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC125307.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC125309.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
BC145157.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
NM_025935.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P425737 |
AK011356.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139864 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125130 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07854288 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P122749 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P117883 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11115112 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11115110 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10798465 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11115116 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988729 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988730 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988731 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28988732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988559 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28988586 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988738 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10806398 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988719 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10806396 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988675 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11040965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988698 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11040964 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988701 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988680 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10806399 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11040959 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988734 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988764 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10529111 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988732 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988684 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988693 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10900288 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10468976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988704 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988612 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11115115 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11115116 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988714 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988597 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988688 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988746 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11083741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988690 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10765903 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10595006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10497847 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988678 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988676 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988767 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988607 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10885108 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988677 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988608 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988712 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988720 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988699 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988694 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10877965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988710 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988755 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988708 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10955609 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988756 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988702 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28988749 |
- |
- |