Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026044.3 | NP_080320.1 | 2 - 3 | 336 | 95 | |
GenBank mRNA | AK013297.1 | - | 2 - 3 | 316 | 95 | |
AK078448.1 | - | 2 - 3 | 299 | 95 | ||
BC086486.1 | - | 2 - 3 | 335 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |