Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026044.3 | NP_080320.1 | 6 - 7 | 770 | 58 | |
GenBank mRNA | AK013297.1 | - | 6 - 7 | 750 | 58 | |
AK078448.1 | - | 6 - 7 | 733 | 58 | ||
AK152000.1 | - | 5 - 6 | 683 | 58 | ||
AK153537.1 | - | 5 - 6 | 683 | 58 | ||
BC056962.1 | - | 4 - 5 | 1420 | 58 | ||
BC086486.1 | - | 6 - 7 | 772 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |