Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026062.4 | NP_080338.1 | 1 - 2 | 132 | 64 | |
GenBank mRNA | AK008504.1 | - | 1 - 2 | 75 | 64 | |
AK013580.1 | - | 1 - 2 | 96 | 64 | ||
AK038811.1 | - | 1 - 2 | 89 | 64 | ||
AK048979.1 | - | 1 - 2 | 89 | 64 | ||
AK050305.1 | - | 1 - 2 | 89 | 64 | ||
AK153488.1 | - | 1 - 2 | 87 | 64 | ||
BC055842.1 | - | 1 - 2 | 64 | 64 | ||
BC061013.1 | - | 1 - 2 | 84 | 64 | ||
BC095935.1 | - | 1 - 2 | 66 | 64 | ||
BC116683.1 | - | 1 - 2 | 77 | 40106565 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |