Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_001122960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_001122960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_026070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_026070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_027487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
NM_027487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008134.mm.1 |
TC1000001075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_001122960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_001122960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_026070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_026070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_027487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
NM_027487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008131.mm.1 |
TC1000001075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_001122960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_001122960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_026070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_026070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_027487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
NM_027487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008132.mm.1 |
TC1000001075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_001122960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_001122960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_026070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_026070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_027487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
NM_027487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008126.mm.1 |
TC1000001075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_001122960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_001122960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_026070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_026070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_027487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
NM_027487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008130.mm.1 |
TC1000001075.mm.1 |
MTA TC |