Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 13 - 14 | 2172 | 86 | |
GenBank mRNA | AK013003.1 | - | 4 - 5 | 389 | 86 | |
AK031153.1 | - | 4 - 5 | 488 | 86 | ||
AK032416.1 | - | 13 - 14 | 2135 | 86 | ||
AK076799.1 | - | 13 - 14 | 1745 | 86 | ||
AK134544.1 | - | 10 - 11 | 1039 | 86 | ||
AK141443.1 | - | 13 - 14 | 1722 | 86 | ||
AK150941.1 | - | 12 - 13 | 1647 | 86 | ||
AK151018.1 | - | 13 - 14 | 2142 | 86 | ||
AK152024.1 | - | 13 - 14 | 2142 | 86 | ||
AK152435.1 | - | 12 - 13 | 1647 | 86 | ||
AK172567.1 | - | 13 - 14 | 2138 | 86 | ||
AK173313.1 | - | 13 - 14 | 1791 | 86 | ||
BC010833.1 | - | 13 - 14 | 1787 | 4235165 | 86 | |
BC054121.1 | - | 13 - 14 | 2158 | 6393910 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |