Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026283.2 | NP_080559.1 | 4 - 5 | 1059 | 66 | |
GenBank mRNA | AK006211.1 | - | 4 - 5 | 1060 | 66 | |
AK015593.1 | - | 4 - 5 | 1037 | 66 | ||
AK075749.1 | - | 4 - 5 | 1010 | 66 | ||
AK147234.1 | - | 4 - 5 | 863 | 66 | ||
AK147528.1 | - | 4 - 5 | 863 | 66 | ||
BC100476.1 | - | 4 - 5 | 1074 | 30916247 | 66 | |
BC116745.1 | - | 4 - 5 | 1047 | 40126064 | 66 | |
BC116747.1 | - | 4 - 5 | 1047 | 40126066 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |