Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026388.2 | NP_080664.1 | 1 - 2 | 333 | 73 | |
GenBank mRNA | AK004777.1 | - | 1 - 2 | 316 | 73 | |
AK155413.1 | - | 1 - 2 | 328 | 73 | ||
AK219916.1 | - | 1 - 2 | 76 | 73 | ||
BC019544.1 | - | 1 - 2 | 297 | 4211201 | 73 | |
BY739996.1 | - | 1 - 2 | 333 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |