Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466591 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466589 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466586 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466587 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466588 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466585 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466590 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
450017 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AJ272504.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK049929.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK146588.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK150388.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK150604.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK151095.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK151501.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK151871.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK161306.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
AK165995.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
BC034161.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
BC043055.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807583 |
NM_019989.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
338645 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750217 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948182 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
91293 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P129276 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466579 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466577 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466586 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466571 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466584 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466580 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466614 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466589 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466572 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466613 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466575 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466587 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466574 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24466582 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05906286 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05906274 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P129276 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P335019 |
AK028830.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK141474.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
BC080818.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK076404.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK079465.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P641758 |
AK028830.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108605 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P129276 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05866624 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05866632 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05866634 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05866633 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05866631 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466591 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466590 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466586 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466585 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466588 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466587 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466584 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24466589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24466596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866633 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05158812 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466562 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05158806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466582 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466613 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866631 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466569 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466559 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05657724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05412221 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866632 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466552 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866634 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466594 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466545 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05412222 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466555 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05866624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466567 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466609 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05172121 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466554 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466610 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466547 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466598 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05821301 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466575 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466563 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05412215 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466612 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05582775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466557 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05649382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466607 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466549 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466568 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24466599 |
- |
- |