Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177576.1 | NP_001171047.1 | 3 - 4 | 366 | 58 | |
NM_026646.3 | NP_080922.1 | 3 - 4 | 608 | 58 | ||
GenBank mRNA | AK010193.1 | - | 3 - 4 | 354 | 58 | |
AK018760.1 | - | 3 - 4 | 372 | 58 | ||
AK046874.1 | - | 3 - 4 | 353 | 58 | ||
AK048389.1 | - | 3 - 4 | 337 | 58 | ||
AK049313.1 | - | 3 - 4 | 337 | 58 | ||
AK155276.1 | - | 3 - 4 | 360 | 58 | ||
AK162400.1 | - | 3 - 4 | 319 | 58 | ||
BC037949.1 | - | 3 - 4 | 365 | 5367108 | 58 | |
BC050887.1 | - | 3 - 4 | 561 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |