Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026647.3 | NP_080923.2 | 6 - 7 | 863 | 87 | |
GenBank mRNA | AK020293.1 | - | 6 - 7 | 745 | 87 | |
AY166674.1 | - | 4 - 5 | 501 | 87 | ||
BC043110.1 | - | 6 - 7 | 709 | 6406930 | 87 | |
BC058372.1 | - | 6 - 7 | 731 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |