Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027117.3 | NP_081393.2 | 8 - 9 | 1089 | 66 | |
GenBank mRNA | AK009481.1 | - | 8 - 9 | 1004 | 66 | |
AK051418.1 | - | 8 - 9 | 1016 | 66 | ||
AY513272.1 | - | 8 - 9 | 1040 | 66 | ||
BC005581.1 | - | 8 - 9 | 1013 | 3968615 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |