Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027222.3 | NP_081498.2 | 2 - 3 | 331 | 89 | |
GenBank mRNA | AK008016.1 | - | 2 - 3 | 332 | 89 | |
AK088094.1 | - | 2 - 3 | 351 | 89 | ||
BC030674.1 | - | 1 - 2 | 113 | 89 | ||
BC064044.1 | - | 2 - 3 | 318 | 875322 | 89 | |
GQ477351.1 | - | 2 - 3 | 297 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |