Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_001122732.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_001122732.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_027029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_027029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_027300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_027300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_028647.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
NM_028647.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006483.mm.1 |
TC0100000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_001122732.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_001122732.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_027029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_027029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_027300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_027300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_028647.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
NM_028647.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006480.mm.1 |
TC0100000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_001122732.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_001122732.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_027029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_027029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_027300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_027300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_028647.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
NM_028647.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006481.mm.1 |
TC0100000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_001122732.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_001122732.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_027029.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_027029.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_027300.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_027300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_028647.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
NM_028647.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006479.mm.1 |
TC0100000777.mm.1 |
MTA TC |