Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027583.3 | NP_081859.1 | 1 - 2 | 108 | 66 | |
NM_029150.4 | NP_083426.1 | 1 - 2 | 108 | 66 | ||
GenBank mRNA | AK014869.1 | - | 1 - 2 | 72 | 66 | |
AK015690.1 | - | 1 - 2 | 77 | 66 | ||
AK076628.1 | - | 1 - 2 | 109 | 66 | ||
BB572700.2 | - | 1 - 2 | 109 | 66 | ||
BC100443.1 | - | 1 - 2 | 89 | 30920480 | 66 | |
BC132205.1 | - | 1 - 2 | 32 | 40130482 | 66 | |
BC132539.1 | - | 1 - 2 | 32 | 40130816 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |