Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027617.3 | NP_081893.1 | 13 - 14 | 1545 | 67 | |
GenBank mRNA | BC139279.1 | - | 12 - 13 | 1458 | 8862301 | 67 |
BC139280.1 | - | 12 - 13 | 1458 | 8862302 | 67 | |
BC171970.1 | - | 12 - 13 | 1456 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |