Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028048.2 | NP_082324.1 | 3 - 4 | 1126 | 81 | |
GenBank mRNA | AK007453.1 | - | 3 - 4 | 1128 | 81 | |
AK035739.1 | - | 1097 | 81 | |||
AK171131.1 | - | 3 - 4 | 1133 | 81 | ||
AK186841.1 | - | 1 - 2 | 82 | 81 | ||
BC004569.1 | - | 3 - 4 | 566 | 81 | ||
BC019996.1 | - | 3 - 4 | 1068 | 4207271 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |