Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028306.3 | NP_082582.1 | 5 - 6 | 573 | 75 | |
GenBank mRNA | AK012548.1 | - | 5 - 6 | 546 | 75 | |
AY196790.1 | - | 4 - 5 | 453 | 75 | ||
BC011103.1 | - | 5 - 6 | 573 | 3990088 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |