Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079814.1 | NP_001073282.1 | 9 - 10 | 862 | 87 | |
NM_028720.2 | NP_082996.2 | 9 - 10 | 862 | 87 | ||
GenBank mRNA | AK014456.1 | - | 9 - 10 | 802 | 87 | |
AK040841.1 | - | 9 - 10 | 812 | 87 | ||
AK140812.1 | - | 7 - 8 | 2460 | 87 | ||
AK145277.1 | - | 9 - 10 | 813 | 87 | ||
AK150349.1 | - | 9 - 10 | 806 | 87 | ||
AK151832.1 | - | 9 - 10 | 809 | 87 | ||
AK152586.1 | - | 9 - 10 | 809 | 87 | ||
AK152887.1 | - | 9 - 10 | 806 | 87 | ||
AK159476.1 | - | 9 - 10 | 816 | 87 | ||
AK188170.1 | - | 4 - 5 | 249 | 87 | ||
BC006893.1 | - | 9 - 10 | 796 | 3599477 | 87 | |
CF174076.1 | - | 4 - 5 | 200 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |