Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028772.3 | NP_083048.1 | 12 - 13 | 2020 | 91 | |
GenBank mRNA | AK004755.1 | - | 12 - 13 | 2021 | 91 | |
BC024126.1 | - | 8 - 9 | 1303 | 91 | ||
BC089599.1 | - | 12 - 13 | 2023 | 30118948 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |