Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500033138.mm.1 |
TC0500003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033138.mm.1 |
NM_028774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033138.mm.1 |
NM_028774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033142.mm.1 |
TC0500003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033142.mm.1 |
NM_028774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033142.mm.1 |
NM_028774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
TC0500003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256086.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256086.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256087.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_001256087.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_028774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033128.mm.1 |
NM_028774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033137.mm.1 |
TC0500003608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033137.mm.1 |
NM_028774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033137.mm.1 |
NM_028774.3 |
Refseq |