Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145827.1 | NP_001139299.1 | 9 - 10 | 1292 | 75 | |
NM_028800.3 | NP_083076.3 | 7 - 8 | 938 | 75 | ||
GenBank mRNA | AB102946.1 | - | 7 - 8 | 769 | 75 | |
AK009154.1 | - | 2 - 3 | 156 | 75 | ||
AK050808.1 | - | 3 - 4 | 241 | 75 | ||
AK154780.1 | - | 9 - 10 | 1283 | 75 | ||
AY336057.1 | - | 7 - 8 | 923 | 75 | ||
BC046981.1 | - | 7 - 8 | 897 | 6478265 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |