Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145827.1 | NP_001139299.1 | 10 - 11 | 1439 | 74 | |
NM_028800.3 | NP_083076.3 | 8 - 9 | 1085 | 74 | ||
GenBank mRNA | AB102946.1 | - | 8 - 9 | 916 | 74 | |
AK009154.1 | - | 3 - 4 | 303 | 74 | ||
AK050808.1 | - | 4 - 5 | 388 | 74 | ||
AK154780.1 | - | 10 - 11 | 1430 | 74 | ||
AY336057.1 | - | 8 - 9 | 1070 | 74 | ||
BC046981.1 | - | 8 - 9 | 1044 | 6478265 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |