Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpcpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042672.1 | NP_001036137.1 | 11 - 12 | 1275 | 115 | |
NM_001291050.1 | NP_001277979.1 | 10 - 11 | 1148 | 115 | ||
NM_001291051.1 | NP_001277980.1 | 10 - 11 | 1178 | 115 | ||
NM_001291052.1 | NP_001277981.1 | 10 - 11 | 1196 | 115 | ||
NM_027096.2 | NP_081372.1 | 9 - 10 | 1072 | 115 | ||
NM_028802.3 | NP_083078.3 | 11 - 12 | 1272 | 115 | ||
GenBank mRNA | AK009563.1 | - | 4 - 5 | 491 | 115 | |
AK030645.1 | - | 11 - 12 | 1226 | 115 | ||
AK031658.1 | - | 10 - 11 | 1151 | 115 | ||
AK049491.1 | - | 3 - 4 | 361 | 115 | ||
AK050318.1 | - | 11 - 12 | 1187 | 115 | ||
AK051728.1 | - | 3 - 4 | 335 | 115 | ||
AK141374.1 | - | 11 - 12 | 1162 | 115 | ||
AK165952.1 | - | 11 - 12 | 1237 | 115 | ||
AK166293.1 | - | 2 - 3 | 206 | 115 | ||
AK166454.1 | - | 10 - 11 | 1161 | 115 | ||
BC006887.1 | - | 10 - 11 | 1120 | 115 | ||
BC033408.1 | - | 9 - 10 | 1002 | 5357683 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |