Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028873.4 | NP_083149.3 | 5 - 6 | 1881 | 96 | |
GenBank mRNA | AK017830.1 | - | 5 - 6 | 1895 | 96 | |
AK032681.1 | - | 5 - 6 | 1863 | 96 | ||
AK045345.1 | - | 5 - 6 | 1938 | 96 | ||
AK045445.1 | - | 5 - 6 | 1975 | 96 | ||
AK082579.1 | - | 5 - 6 | 1882 | 96 | ||
AK083878.1 | - | 5 - 6 | 1893 | 96 | ||
AK159388.1 | - | 5 - 6 | 1894 | 96 | ||
AK168935.1 | - | 5 - 6 | 1973 | 96 | ||
AK219646.1 | - | 1 - 2 | 146 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |