Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028903.2 | NP_083179.2 | 7 - 8 | 1495 | 70 | |
GenBank mRNA | AK133176.1 | - | 7 - 8 | 1496 | 70 | |
BC016096.1 | - | 7 - 8 | 1471 | 4506472 | 70 | |
BC023907.1 | - | 7 - 8 | 1411 | 5321978 | 70 | |
BC026758.1 | - | 4 - 5 | 521 | 70 | ||
BC032159.1 | - | 7 - 8 | 1451 | 5354657 | 70 | |
DQ122126.1 | - | 6 - 7 | 1145 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |