Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028959.3 | NP_083235.3 | 5 - 6 | 717 | 64 | |
GenBank mRNA | AK017134.1 | - | 5 - 6 | 709 | 64 | |
AK034892.1 | - | 5 - 6 | 710 | 64 | ||
AK161033.1 | - | 1 - 2 | 42 | 64 | ||
BC080681.1 | - | 5 - 6 | 673 | 64 | ||
BC100549.1 | - | 4 - 5 | 567 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |