Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290148.1 | NP_001277077.1 | 5 - 6 | 493 | 78 | |
NM_028964.4 | NP_083240.2 | 12 - 13 | 1542 | 78 | ||
GenBank mRNA | AF399755.1 | - | 5 - 6 | 448 | 78 | |
AK017102.1 | - | 5 - 6 | 494 | 78 | ||
AK162238.1 | - | 4 - 5 | 371 | 78 | ||
BB623950.1 | - | 4 - 5 | 371 | 78 | ||
BC034114.1 | - | 2 - 3 | 142 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |