Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285452.1 | NP_001272381.1 | 1 - 2 | 463 | 99 | |
NM_028975.4 | NP_083251.2 | 2 - 3 | 306 | 99 | ||
NM_030108.2 | NP_084384.1 | 2 - 3 | 306 | 99 | ||
GenBank mRNA | AK005493.1 | - | 2 - 3 | 198 | 99 | |
AK007607.1 | - | 2 - 3 | 114 | 99 | ||
AK007750.1 | - | 2 - 3 | 195 | 99 | ||
AK012413.1 | - | 2 - 3 | 220 | 99 | ||
AK017281.1 | - | 2 - 3 | 175 | 99 | ||
AK019127.1 | - | 1 - 2 | 132 | 99 | ||
AK032555.1 | - | 2 - 3 | 213 | 99 | ||
AK153441.1 | - | 2 - 3 | 131 | 99 | ||
AK165369.1 | - | 2 - 3 | 108 | 99 | ||
BC005562.1 | - | 2 - 3 | 107 | 3965864 | 99 | |
BC016570.1 | - | 1 - 2 | 179 | 3583937 | 99 | |
BC023966.1 | - | 1 - 2 | 172 | 5342147 | 99 | |
CJ103460.1 | - | 1 - 2 | 224 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |