Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027448.2 | NP_081724.1 | 6 - 7 | 1466 | 105 | |
NM_029434.3 | NP_083710.2 | 6 - 7 | 1362 | 105 | ||
GenBank mRNA | AK015260.1 | - | 6 - 7 | 1334 | 105 | |
BC049101.1 | - | 6 - 7 | 1394 | 5702365 | 105 | |
BC052060.1 | - | 6 - 7 | 1394 | 5702365 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |