Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029522.2 | NP_083798.2 | 7 - 8 | 1303 | 79 | |
GenBank mRNA | AK040996.1 | - | 7 - 8 | 1192 | 79 | |
AK143552.1 | - | 7 - 8 | 1304 | 79 | ||
AY081187.1 | - | 7 - 8 | 1147 | 79 | ||
BC021308.1 | - | 6 - 7 | 858 | 5007832 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |