Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161628.1 | NP_001155100.1 | 9 - 10 | 1486 | 67 | |
NM_001161629.1 | NP_001155101.1 | 7 - 8 | 974 | 67 | ||
NM_029946.4 | NP_084222.4 | 26 - 27 | 3593 | 67 | ||
GenBank mRNA | AK016516.1 | - | 7 - 8 | 1079 | 67 | |
AK019850.1 | - | 9 - 10 | 1486 | 67 | ||
AK163363.1 | - | 4 - 5 | 664 | 67 | ||
AK220412.1 | - | 9 - 10 | 1405 | 67 | ||
BC056955.1 | - | 14 - 15 | 1996 | 6812632 | 67 | |
BC065111.1 | - | 26 - 27 | 3593 | 30536704 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |