Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142728.1 | NP_001136200.1 | 4 - 5 | 740 | 87 | |
NM_001142729.1 | NP_001136201.1 | 4 - 5 | 659 | 87 | ||
NM_030070.3 | NP_084346.2 | 4 - 5 | 740 | 87 | ||
GenBank mRNA | AK015292.1 | - | 4 - 5 | 658 | 87 | |
AK019545.1 | - | 4 - 5 | 673 | 87 | ||
AK029802.1 | - | 4 - 5 | 741 | 87 | ||
AK031279.1 | - | 4 - 5 | 659 | 87 | ||
AK039037.1 | - | 4 - 5 | 550 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |